Thermo Scientific Chemicals

Sodium cyanide, 98+%, extra pure, Thermo Scientific Chemicals

Catalog number: 370310010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Sodium cyanide, 98+%, extra pure, Thermo Scientific Chemicals

Catalog number: 370310010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 370310010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
143-33-9
IUPAC Name
sodium iminomethanide
Molecular Formula
CNNa
InChI Key
MNWBNISUBARLIT-UHFFFAOYSA-N
SMILES
[Na+].[C-]#N
Sodium hydroxide (NaOH)
0.3 to 0.8 %
Sodium formate
0.1 to 0.5 %
Appearance (Color)
White
Titration Argentometric
>=98.0 %
Sodium carbonate (Na2CO3)
0.2 to 0.8 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text