Sodium cyanide, 98+%, extra pure, Thermo Scientific Chemicals
Sodium cyanide, 98+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium cyanide, 98+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 kg
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 370310050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS143-33-9
IUPAC Namesodium iminomethanide
Molecular FormulaCNNa
InChI KeyMNWBNISUBARLIT-UHFFFAOYSA-N
SMILES[Na+].[C-]#N
View more
SpecificationsSpecification SheetSpecification Sheet
Sodium hydroxide (NaOH)0.3 to 0.8 %
Sodium formate0.1 to 0.5 %
Appearance (Color)White
Titration Argentometric>=98.0 %
Sodium carbonate (Na2CO3)0.2 to 0.8 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only