4-Mercaptophenol, 99%, Thermo Scientific Chemicals
4-Mercaptophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Mercaptophenol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 370630050
Price (USD)/ Each
51.65
Online exclusive
60.20 
Save 8.55 (14%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
51.65
Online exclusive
60.20 
Save 8.55 (14%)
Add to cart
4-Mercaptophenol, 99%, Thermo Scientific Chemicals
Catalog number370630050
Price (USD)/ Each
51.65
Online exclusive
60.20 
Save 8.55 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-sulfanylphenol
Molecular FormulaC6H6OS
InChI KeyBXAVKNRWVKUTLY-UHFFFAOYSA-N
SMILESOC1=CC=C(S)C=C1
Molecular Weight (g/mol)126.17
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid or low melting solid
GC>=97.5 %
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only