Thermo Scientific Chemicals

Phenyltrimethoxysilane, 85%, Thermo Scientific Chemicals

Catalog number: 370642500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Phenyltrimethoxysilane, 85%, Thermo Scientific Chemicals

Catalog number: 370642500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 370642500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2996-92-1
IUPAC Name
trimethoxy(phenyl)silane
Molecular Formula
C9H14O3Si
InChI Key
ZNOCGWVLWPVKAO-UHFFFAOYSA-N
SMILES
CO[Si](OC)(OC)C1=CC=CC=C1
Refractive index
1.4730 to 1.4750 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=80 %
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text