Thermo Scientific Chemicals

(R)-1-(4-Chlorophenyl)ethylamine, 97%, 98% ee, Thermo Scientific Chemicals

Catalog number: 370650250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

(R)-1-(4-Chlorophenyl)ethylamine, 97%, 98% ee, Thermo Scientific Chemicals

Catalog number: 370650250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 370650250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
27298-99-3
IUPAC Name
1-(4-chlorophenyl)ethan-1-amine
Molecular Formula
C8H10ClN
InChI Key
PINPOEWMCLFRRB-UHFFFAOYNA-N
SMILES
CC(N)C1=CC=C(Cl)C=C1
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to pale yellow
Enantiomeric excess
>=97.5 %
Infrared spectrum
Conforms
Refractive index
1.5400 to 1.5440 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text