Thermo Scientific™

SuperSignal™ ELISA Pico Chemiluminescent Substrate

Catalog number: 37070
Have Questions?
Thermo Scientific™

SuperSignal™ ELISA Pico Chemiluminescent Substrate

Catalog number: 37070
Have Questions?
Catalog Number
37070
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
37070100 mL
-
37069250 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific SuperSignal ELISA Pico Chemiluminescent Substrate is optimized to generate an intense light signal and provide exceptional performance in luminometer-based assays.

Features of SuperSignal ELISA Pico Chemiluminescent Substrate:

Immediate light generation—intense signal is produced immediately at room temperature or at 37°C; emits light at 425nm
High signal:noise ratio—minimal background
Low picogram sensitivity—detects protein in ELISAs down to the picogram levels
Room temperature storage—product with ambient shipping and room temperature storage conditions
8-hour working solution stability—consistent performance of the working solution over an 8-hour period with only a 10% decrease in activity at 24 hours
Flexible—signal can be read in black or white opaque plates

SuperSignal ELISA Pico Chemiluminescent Substrate offers greater sensitivity in ELISAs or any other solution-based assay, using chemiluminescent substrates for enzyme detection and quantification. These ELISAs can take place in either a test tube or a microplate and are quantified by measuring relative light units (RLU) in a luminometer.

Chemiluminescent substrates differ from other substrates in that the light detected is a transient product of the reaction that is only present while the enzyme-substrate reaction is occurring. This is in contrast to substrates that produce a stable, colored product; these colors persist in the well after the enzyme-substrate reaction has terminated.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
ELISA Substrate
Quantity
100 mL Kit
Detection Method
Chemiluminescent
Product Line
SuperSignal™
Substrate Properties
Chemical Substrate
Substrate Type
HRP (Horseradish Peroxidase)
Kit Contents
Stable Peroxide Buffer, 50mL, Luminol/Enhancer, 50mL

Contents & Storage

Upon receipt store product at room temperature. Product shipped at ambient temperature.Introduction

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text