Thermo Scientific Chemicals

Piperonyloyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 370790010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Piperonyloyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 370790010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 370790010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25054-53-9
IUPAC Name
2H-1,3-benzodioxole-5-carbonyl chloride
Molecular Formula
C8H5ClO3
InChI Key
ZRSGZIMDIHBXIN-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=C2OCOC2=C1
Infrared spectrum
Conforms
Appearance (Color)
White to yellow to light beige
Appearance (Form)
Crystalline powder or crystals and/or chunks
Melting point
77°C to 81°C
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text