2-Bromo-5-nitroanisole, 98%, Thermo Scientific Chemicals
2-Bromo-5-nitroanisole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-5-nitroanisole, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
Catalog number 370800100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS77337-82-7
IUPAC Name1-bromo-2-methoxy-4-nitrobenzene
Molecular FormulaC7H6BrNO3
InChI KeyNTKADLOYTKVXQN-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1Br)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light brown
Melting point99°C to 103°C
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only