Chemical Identifiers
CAS475-25-2
IUPAC Name5,6,10,15-tetrahydroxy-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-1(17),2,4,6,12,15-hexaen-14-one
Molecular FormulaC16H12O6
InChI KeyHLUCICHZHWJHLL-UHFFFAOYNA-N
SMILESOC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O
View more