Thermo Scientific Chemicals

Ethyl benzoylacetate, 90%, Thermo Scientific Chemicals

Catalog number: 370850500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl benzoylacetate, 90%, Thermo Scientific Chemicals

Catalog number: 370850500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 370850500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
94-02-0
IUPAC Name
ethyl 3-oxo-3-phenylpropanoate
Molecular Formula
C11H12O3
InChI Key
GKKZMYDNDDMXSE-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(=O)C1=CC=CC=C1
Refractive index
1.5240 to 1.5330 (20°C, 589 nm)
HPLC
>=88 %
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to yellow to red to brown
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text