Thermo Scientific Chemicals

Ethylboronic acid, 95%, Thermo Scientific Chemicals

Catalog number: 370860010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethylboronic acid, 95%, Thermo Scientific Chemicals

Catalog number: 370860010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 370860010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4433-63-0
IUPAC Name
ethylboronic acid
Molecular Formula
C2H7BO2
InChI Key
PAVZHTXVORCEHP-UHFFFAOYSA-N
SMILES
CCB(O)O
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder or flakes or needles
Infrared spectrum
Conforms
Titration with NaOH
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text