Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 370911000
Price (USD)/ Each
56.65
Online exclusive
66.90 
Save 10.25 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
56.65
Online exclusive
66.90 
Save 10.25 (15%)
Add to cart
Phenylacetaldehyde, 98%, stabilized, Thermo Scientific Chemicals
Catalog number370911000
Price (USD)/ Each
56.65
Online exclusive
66.90 
Save 10.25 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-phenylacetaldehyde
Molecular FormulaC8H8O
InChI KeyDTUQWGWMVIHBKE-UHFFFAOYSA-N
SMILESO=CCC1=CC=CC=C1
Molecular Weight (g/mol)120.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5250 to 1.5350 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only