Ammonium bicarbonate, 98%, pure
Ammonium bicarbonate, 98%, pure
Ammonium bicarbonate, 98%, pure
Ammonium bicarbonate, 98%, pure
Thermo Scientific Chemicals

Ammonium bicarbonate, 98%, pure

CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 370930010
Price (USD)
89.80
Each
In stock
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
89.80
Each
Add to cart
Chemical Identifiers
CAS1066-33-7
IUPAC Namecarbonic acid amine
Molecular FormulaCH5NO3
InChI KeyATRRKUHOCOJYRX-UHFFFAOYSA-N
SMILESN.OC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HCl>=97.5 %
Anticaking agent=<0.55 % (MgCO3)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection