Ammonium bicarbonate, 98%, pure, Thermo Scientific Chemicals
Ammonium bicarbonate, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium bicarbonate, 98%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 370930250
Price (USD)/ Each
35.65
Online exclusive
38.90 
Save 3.25 (8%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
35.65
Online exclusive
38.90 
Save 3.25 (8%)
Add to cart
Ammonium bicarbonate, 98%, pure, Thermo Scientific Chemicals
Catalog number370930250
Price (USD)/ Each
35.65
Online exclusive
38.90 
Save 3.25 (8%)
-
Add to cart
Chemical Identifiers
CAS1066-33-7
IUPAC Namedizinc(2+) diammonium tricarbonate
Molecular FormulaC3H8N2O9Zn2
InChI KeyQOPSSYZLVIENBR-UHFFFAOYSA-J
SMILES[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HCl>=97.5 %
Anticaking agent=<0.55 % (MgCO3)
Appearance (Form)Crystalline powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only