Thermo Scientific Chemicals

Sand, pure, 40-100 mesh, Thermo Scientific Chemicals

Catalog number: 370940250
25 kg, Fiber Drum, Each
Thermo Scientific Chemicals

Sand, pure, 40-100 mesh, Thermo Scientific Chemicals

Catalog number: 370940250
25 kg, Fiber Drum, Each
Quantity
Packaging
Catalog number: 370940250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14808-60-7
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Appearance (Color)
Light beige to grey
Appearance (Form)
Crystals
Particle size
40 to 120 mesh
Chromium(III) oxide (Cr2O3)
0.0002 %
Particle size
207 µm (AGS) Typical

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text