Thermo Scientific Chemicals

tert-Butyl acrylate, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 371130051
5 L, Plastic drum, Each
Thermo Scientific Chemicals

tert-Butyl acrylate, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 371130051
5 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 371130051
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1663-39-4
IUPAC Name
tert-butyl prop-2-enoate
Molecular Formula
C7H12O2
InChI Key
ISXSCDLOGDJUNJ-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)C=C
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Form)
Clear liquid
Color scale
=<10 APHA
Refractive index
1.4100 to 1.4120 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text