tert-Butyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
tert-Butyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl acrylate, 99%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 371131000
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.20 
Save 8.55 (14%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.20 
Save 8.55 (14%)
Add to cart
tert-Butyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
Catalog number371131000
Price (USD)/ Each
52.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
61.20 
Save 8.55 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Nametert-butyl prop-2-enoate
Molecular FormulaC7H12O2
InChI KeyISXSCDLOGDJUNJ-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)C=C
Molecular Weight (g/mol)128.17
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Clear liquid
Color scale=<10 APHA
Refractive index1.4100 to 1.4120 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only