Thermo Scientific Chemicals

1,8-Diaminonaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: 371181000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

1,8-Diaminonaphthalene, 97%, Thermo Scientific Chemicals

Catalog number: 371181000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 371181000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
479-27-6
IUPAC Name
naphthalene-1,8-diamine
Molecular Formula
C10H10N2
InChI Key
YFOOEYJGMMJJLS-UHFFFAOYSA-N
SMILES
NC1=CC=CC2=CC=CC(N)=C12
Appearance (Color)
Grey to red to purple to dark brown
Appearance (Form)
Crystals or flakes and/or chunks
Infrared spectrum
Conforms
Melting point
61°C to 66°C
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text