Chemical Identifiers
CAS1306-06-5
IUPAC Namepentacalcium hydroxide triphosphate
Molecular FormulaCa5HO13P3
InChI KeyXYJRXVWERLGGKC-UHFFFAOYSA-D
SMILES[OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more