2,3-Dichlorophenethylamine, 99+%, Thermo Scientific Chemicals
2,3-Dichlorophenethylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dichlorophenethylamine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 371290050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS34164-43-7
IUPAC Name2-(2,3-dichlorophenyl)ethan-1-aminium
Molecular FormulaC8H10Cl2N
InChI KeyWWRLSNWKOZHYIY-UHFFFAOYSA-O
SMILES[NH3+]CCC1=CC=CC(Cl)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4690 to 1.4710 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only