Thermo Scientific Chemicals

(Triphenylphosphoranylidene)acetonitrile, 96%, Thermo Scientific Chemicals

Catalog number: 371570050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

(Triphenylphosphoranylidene)acetonitrile, 96%, Thermo Scientific Chemicals

Catalog number: 371570050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 371570050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16640-68-9
IUPAC Name
2-(triphenyl-λ⁵-phosphanylidene)acetonitrile
Molecular Formula
C20H16NP
InChI Key
APISVOVOJVZIBA-UHFFFAOYSA-N
SMILES
N#CC=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Infrared spectrum
Conforms
Additional info
This product unavoidably and progressively
Appearance (Color)
Off-white to light yellow
Appearance (Form)
Crystalline powder
Additional info
The actual assay is mentioned on the COA.

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text