Thermo Scientific Chemicals

L-Asparagine, 99%, Thermo Scientific Chemicals

Catalog number: 371601000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

L-Asparagine, 99%, Thermo Scientific Chemicals

Catalog number: 371601000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 371601000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
70-47-3
IUPAC Name
2-amino-3-carbamoylpropanoic acid
Molecular Formula
C4H8N2O3
InChI Key
DCXYFEDJOCDNAF-UHFFFAOYNA-N
SMILES
NC(CC(N)=O)C(O)=O
Titration with HClO4
>=98.5 % (On dry substance)
Infrared spectrum
Conforms
Appearance (Color)
White
Water
=<1 %
Specific optical rotation
+33° to +36° (20°C, 589 nm) (c=10, 2N HCl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text