Pinacolborane, 97%, stabilized, Thermo Scientific Chemicals
Pinacolborane, 97%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pinacolborane, 97%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 371630250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC6H13BO2
InChI KeyUCFSYHMCKWNKAH-UHFFFAOYSA-N
SMILESCC1(C)OBOC1(C)C
Molecular Weight (g/mol)127.98
View more
SpecificationsSpecification SheetSpecification Sheet
Stabilizer0.1 to 1.5 % Triethylamine
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.3950 to 1.4000 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only