Thermo Scientific Chemicals

Benzyl 3-pyrroline-1-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: 371690050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

Benzyl 3-pyrroline-1-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: 371690050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 371690050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
31970-04-4
IUPAC Name
benzyl 2,5-dihydro-1H-pyrrole-1-carboxylate
Molecular Formula
C12H13NO2
InChI Key
XSKKIFJNZPNVGO-UHFFFAOYSA-N
SMILES
O=C(OCC1=CC=CC=C1)N1CC=CC1
Appearance (Color)
Colorless to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text