2-Fluoro-5-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals
2-Fluoro-5-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoro-5-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Plastic bottle
Catalog number 371730010
Price (USD)/ Each
78.70
-
Add to cart
Quantity:
1 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
78.70
Add to cart
2-Fluoro-5-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals
Catalog number371730010
Price (USD)/ Each
78.70
-
Add to cart
Chemical Identifiers
CAS27996-87-8
IUPAC Name2-fluoro-5-nitrobenzaldehyde
Molecular FormulaC7H4FNO3
InChI KeyVVXFDFQEIRGULC-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(F)C(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point56°C to 60°C
Appearance (Color)Beige to light brown
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only