Thermo Scientific Chemicals

3-Biphenylcarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 371770010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Biphenylcarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: 371770010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 371770010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
716-76-7
IUPAC Name
[1,1'-biphenyl]-3-carboxylic acid
Molecular Formula
C13H10O2
InChI Key
XNLWJFYYOIRPIO-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC(=C1)C1=CC=CC=C1
Appearance (Form)
Crystalline powder
Titration with NaOH
>=96.0 %
Appearance (Color)
Off-white to beige
Infrared spectrum
Conforms
Melting point
164°C to 169°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text