Thermo Scientific Chemicals

Methyl acetate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 371830025
2.5 L, Each
Thermo Scientific Chemicals

Methyl acetate, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 371830025
2.5 L, Each
Quantity
Catalog number: 371830025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
79-20-9
IUPAC Name
methyl acetate
Molecular Formula
C3H6O2
InChI Key
KXKVLQRXCPHEJC-UHFFFAOYSA-N
SMILES
COC(C)=O
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Water
=<0.005 %
GC
>=99.0 %
Residue after evaporation
=<0.002 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text