cis-2,6-Dimethylmorpholine, 97%, Thermo Scientific Chemicals
cis-2,6-Dimethylmorpholine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-2,6-Dimethylmorpholine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 371841000
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
Add to cart
cis-2,6-Dimethylmorpholine, 97%, Thermo Scientific Chemicals
Catalog number371841000
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
CAS6485-55-8
IUPAC Name(2R,6S)-2,6-dimethylmorpholin-4-ium
Molecular FormulaC6H14NO
InChI KeyHNVIQLPOGUDBSU-OLQVQODUSA-O
SMILESC[C@H]1C[NH2+]C[C@@H](C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.4450 to 1.4470 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only