Methyl 3-(dimethylamino)-2-isocyanoacrylate, 96%, Thermo Scientific Chemicals
Methyl 3-(dimethylamino)-2-isocyanoacrylate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3-(dimethylamino)-2-isocyanoacrylate, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 372020010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS113212-14-9
IUPAC Namemethyl (2Z)-3-(dimethylamino)-2-isocyanoprop-2-enoate
Molecular FormulaC7H10N2O2
InChI KeyFREGRQYTSPAJDX-WAYWQWQTSA-N
SMILESCOC(=O)C(=C\N(C)C)\[N+]#[C-]
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point61°C to 67°C
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)Yellow
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only