Thermo Scientific Chemicals

cis-Epoxysuccinic acid, 97%, Thermo Scientific Chemicals

Catalog number: 372640050
5 g, Each
Thermo Scientific Chemicals

cis-Epoxysuccinic acid, 97%, Thermo Scientific Chemicals

Catalog number: 372640050
5 g, Each
Quantity
Catalog number: 372640050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16533-72-5
IUPAC Name
(2R,3S)-oxirane-2,3-dicarboxylate
Molecular Formula
C4H2O5
InChI Key
DCEMCPAKSGRHCN-XIXRPRMCSA-L
SMILES
[O-]C(=O)[C@H]1O[C@H]1C([O-])=O
Titration with NaOH
96.0 to 104.0 %
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text