2,3-Dihydro-6-isocyano-1,4-benzodioxine, 95%, Thermo Scientific Chemicals
2,3-Dihydro-6-isocyano-1,4-benzodioxine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,3-Dihydro-6-isocyano-1,4-benzodioxine, 95%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 372770010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name6-isocyano-2,3-dihydro-1,4-benzodioxine
Molecular FormulaC9H7NO2
InChI KeyAAUQOWLQNFIDQL-UHFFFAOYSA-N
SMILES[C-]#[N+]C1=CC2=C(OCCO2)C=C1
Molecular Weight (g/mol)161.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Appearance (Form)Powder or crystals and/or chunks
Melting point81°C to 87°C
HPLC>=94 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only