Thermo Scientific Chemicals

4-Hydroxy-6-methylcoumarin, 98+%, Thermo Scientific Chemicals

Catalog number: 372870050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Hydroxy-6-methylcoumarin, 98+%, Thermo Scientific Chemicals

Catalog number: 372870050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 372870050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13252-83-0
IUPAC Name
2-hydroxy-6-methyl-4H-chromen-4-one
Molecular Formula
C10H8O3
InChI Key
CSVFZDYJGBVNEX-UHFFFAOYSA-N
SMILES
CC1=CC=C2OC(O)=CC(=O)C2=C1
Titration with TBAH
>=98.0 %
Infrared spectrum
Conforms
Appearance (Form)
Powder
Melting point
258°C to 262°C
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text