1x1 image pixel for data collection
Mexiletine hydrochloride
Mexiletine hydrochloride
Mexiletine hydrochloride
Mexiletine hydrochloride
Thermo Scientific Chemicals

Mexiletine hydrochloride

CAS: 5370-01-4 | C11H17NO·HCl | 215.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 372910250
Price (USD)
66.65
Online Exclusive
73.90
Save 7.25 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
73.90
Save 7.25 (10%)
Each
Add to cart
Chemical Identifiers
CAS5370-01-4
IUPAC Namehydrogen 1-(2,6-dimethylphenoxy)propan-2-amine chloride
Molecular FormulaC11H18ClNO
InChI KeyNFEIBWMZVIVJLQ-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(N)COC1=C(C)C=CC=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point200°C to 205°C
Loss on drying=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link