3731-05-RIPK also known as 3731-05-RI
Full specifications
Pack of 960 300L, Non-Filtered, Sterile, Refill 300 μL Orange - Description 300L, Non-Filtered, Sterile, Refill Volume (Metric) 300 μL Filtered Non-filtered Sterility Sterile Color Natural Model 300L Length (English) 2.26 in. Length (Metric) 5.75cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Orange For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Pack of 960
3722-05-RI Full specifications
Case of 4800 20L, Non-Filtered, Non-sterile, Refill 20 μL Red 388.85 Online price 707.00
- Description 20L, Non-Filtered, Non-sterile, Refill Volume (Metric) 20 μL Filtered Non-filtered Sterility Non-sterile Color Natural Model 20L Length (English) 1.95 in. Length (Metric) 4.86cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Red For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Case of 4800
3722-05-RIPK also known as 3722-05-RI
Full specifications
Pack of 960 20L, Non-Filtered, Non-sterile, Refill 20 μL Red - Description 20L, Non-Filtered, Non-sterile, Refill Volume (Metric) 20 μL Filtered Non-filtered Sterility Non-sterile Color Natural Model 20L Length (English) 1.95 in. Length (Metric) 4.86cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Red For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Pack of 960
3752-05-RI Full specifications
Case of 4800 200L, Non-Filtered, Non-sterile, Refill 200 μL Green 391.60 Online price 712.00
- Description 200L, Non-Filtered, Non-sterile, Refill Volume (Metric) 200 μL Filtered Non-filtered Sterility Non-sterile Color Natural Model 200L Length (English) 1.97 in. Length (Metric) 5.00cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Green For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Case of 4800
3752-05-RIPK also known as RI3752-05-
Full specifications
Pack of 960 200L, Non-Filtered, Non-sterile, Refill 200 μL Green - Description 200L, Non-Filtered, Non-sterile, Refill Volume (Metric) 200 μL Filtered Non-filtered Sterility Non-sterile Color Natural Model 200L Length (English) 1.97 in. Length (Metric) 5.00cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Green For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Pack of 960
3732-05-RI Full specifications
Case of 4800 300L, Non-Filtered, Non-sterile, Refill 300 μL Orange - Description 300L, Non-Filtered, Non-sterile, Refill Volume (Metric) 300 μL Filtered Non-filtered Sterility Non-sterile Color Natural Model 300L Length (English) 2.26 in. Length (Metric) 5.75cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Orange For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Case of 4800
3732-05-RIPK also known as 3732-05-RI
Full specifications
Pack of 960 300L, Non-Filtered, Non-sterile, Refill 300 μL Orange - Description 300L, Non-Filtered, Non-sterile, Refill Volume (Metric) 300 μL Filtered Non-filtered Sterility Non-sterile Color Natural Model 300L Length (English) 2.26 in. Length (Metric) 5.75cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Orange For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Pack of 960
3782-05-RI Full specifications
Case of 3072 1000L, Non-Filtered, Non-sterile, Refill 1000 μL Blue 397.19 Online price 531.00
- Description 1000L, Non-Filtered, Non-sterile, Refill Volume (Metric) 1000 μL Filtered Non-filtered Sterility Non-sterile Color Natural Model 1000L Length (English) 3.39 in. Length (Metric) 8.61cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Blue For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Case of 3072
3782-05-RIPK also known as 3782-05-RI
Full specifications
Each 1000L, Non-Filtered, Non-sterile, Refill 1000 μL Blue - Description 1000L, Non-Filtered, Non-sterile, Refill Volume (Metric) 1000 μL Filtered Non-filtered Sterility Non-sterile Color Natural Model 1000L Length (English) 3.39 in. Length (Metric) 8.61cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Blue For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Each
3742-05-RI Full specifications
Case of 3072 1200L, Non-Filtered, Non-sterile, Refill 1200 μL Yellow Contact Us
- - Description 1200L, Non-Filtered, Non-sterile, Refill Volume (Metric) 1200 μL Filtered Non-filtered Sterility Non-sterile Color Natural Model 1200L Length (English) 3.42 in. Length (Metric) 8.67cm Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free Format Refill Tip Style Universal Color Code Yellow For Use With (Equipment) Rainin LTS LiteTouch Product Line ART SoftFit L Unit Size Case of 3072