Potassium iodide, 99+%, for analysis, Thermo Scientific Chemicals
Potassium iodide, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium iodide, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 373650025
Price (USD)/ Each
599.65
Online exclusive
705.00 
Save 105.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
599.65
Online exclusive
705.00 
Save 105.35 (15%)
Add to cart
Potassium iodide, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number373650025
Price (USD)/ Each
599.65
Online exclusive
705.00 
Save 105.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
Molecular Weight (g/mol)166.00
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Titration Argentometric99.0 to 101.0 % (on dry substance)
Iron (Fe)=<5 ppm
Appearance (Form)Crystalline powder or crystals
Alkalinity=<0.022 % (as KOH)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only