Thermo Scientific Chemicals

Potassium iodide, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 373655000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium iodide, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 373655000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 373655000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7681-11-0
IUPAC Name
potassium iodide
Molecular Formula
IK
InChI Key
NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES
[K+].[I-]
Appearance (Color)
Colorless to white
Titration Argentometric
99.0 to 101.0 % (on dry substance)
Iron (Fe)
=<5 ppm
Appearance (Form)
Crystalline powder or crystals
Alkalinity
=<0.022 % (as KOH)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text