Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Nunc Cryobank and Bank-it Rack, Tube Press out Tool, 1 Each |
Type | Tube Press Out Tool |
Vial Type | Cryobank |
Color | Silver |
No. per Pack | 1 |
No. per Case | 1 |
For Use With (Equipment) | Cryobank and Bank-It Systems |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
374009 | Each | Nunc Cryobank and Bank-it Rack, Tube Press out Tool, 1 Each | 1,746.00 Online price 2,216.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|