1x1 image pixel for data collection
Methyl 3-methylphenylacetate, 98%
Methyl 3-methylphenylacetate, 98%
Methyl 3-methylphenylacetate, 98%
Methyl 3-methylphenylacetate, 98%
Thermo Scientific Chemicals

Methyl 3-methylphenylacetate, 98%

CAS: 53088-69-0 | C10H12O2 | 164.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 374080250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS53088-69-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5045 to 1.5065 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link