Search Thermo Fisher Scientific
Catalog Number | Closure Color | Description | Packaging | Fit for Purpose |
---|---|---|---|---|
3743 | Colorless | V-Bottom ScrewTop Tubes | Bulk, 480 Tubes | Sterile & PCR Ready |
3743RED | Red | V-Bottom ScrewTop Tubes | Bulk, 480 Tubes | Sterile & PCR Ready |
3743YEL | Yellow | V-Bottom ScrewTop Tubes | Bulk, 480 Tubes | Sterile & PCR Ready |
3743BLU | Blue | V-Bottom ScrewTop Tubes | Bulk, 480 Tubes | Sterile & PCR Ready |
3743GRE | Green | V-Bottom ScrewTop Tubes | Bulk, 480 Tubes | Sterile & PCR Ready |
3743WHI | White | V-Bottom ScrewTop Tubes | Bulk, 480 Tubes | Sterile & PCR Ready |
3743PUR | Purple | V-Bottom ScrewTop Tubes | Bulk, 480 Tubes | Sterile & PCR Ready |
3744 | Colorless | V-Bottom ScrewTop Tubes | 5 Latch Racks of 96 Tubes | Sterile & PCR Ready |
3744RED | Red | V-Bottom ScrewTop Tubes | 5 Latch Racks of 96 Tubes | Sterile & PCR Ready |
3744YEL | Yellow | V-Bottom ScrewTop Tubes | 5 Latch Racks of 96 Tubes | Sterile & PCR Ready |
3744BLU | Blue | V-Bottom ScrewTop Tubes | 5 Latch Racks of 96 Tubes | Sterile & PCR Ready |
3744GRE | Green | V-Bottom ScrewTop Tubes | 5 Latch Racks of 96 Tubes | Sterile & PCR Ready |
3744WHI | White | V-Bottom ScrewTop Tubes | 5 Latch Racks of 96 Tubes | Sterile & PCR Ready |
3744PUR | Purple | V-Bottom ScrewTop Tubes | 5 Latch Racks of 96 Tubes | Sterile & PCR Ready |
3745 | n.a. | V-Bottom ScrewTop Tubes, No Caps | 5 Latch Racks of 96 Tubes | Sterile & PCR Ready |
3744AMB | n.a. | 2D tube with no cap | 5 Latch Racks of 96 Tubes | Sterile & PCR Ready |
3743AMB | Red | 2D tube with red cap | 5 Latch Racks of 96 Tubes | Sterile & PCR Ready |
3744-WP | Colorless | V-Bottom ScrewTop Tubes with white patch | 5 Latch Racks of 96 Tubes | Sterile & PCR Ready |
Store samples securely at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 500μL ScrewTop tubes. These tubes are molded from medical grade resins and feature a permanent, high contrast 2D barcode. Used in combination with Thermo Scientific™ VisionMate™ 2D barcode readers, manual and automated capping equipment, and labeling options, the 2D ScrewTop Tubes meet a range of manual and automated long term storage needs.