2-Iodoxybenzoic acid, stabilized, Thermo Scientific Chemicals
2-Iodoxybenzoic acid, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Iodoxybenzoic acid, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 374650100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-hydroxy-1-oxo-3H-1λ⁵,2-benziodaoxol-3-one
Molecular FormulaC7H5IO4
InChI KeyCQMJEZQEVXQEJB-UHFFFAOYSA-N
SMILESO[I]1(=O)OC(=O)C2=CC=CC=C12
Molecular Weight (g/mol)280.02
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Powder or crystals
IdentificationIsophthalic acid, Benzoic acid)
Titration Iodometric39 to 50 % as 2-Iodoxybenzoic acid
IdentificationHPLC (2-Iodoxybenzoic acid, 2-Iodosobenzoic acid,
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagent
RUO – Research Use Only