Thermo Scientific Chemicals

Triethyl citrate, 99%, Thermo Scientific Chemicals

Catalog number: 375062500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Triethyl citrate, 99%, Thermo Scientific Chemicals

Catalog number: 375062500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 375062500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-93-0
IUPAC Name
1,2,3-triethyl 2-hydroxypropane-1,2,3-tricarboxylate
Molecular Formula
C12H20O7
InChI Key
DOOTYTYQINUNNV-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC
Appearance (Form)
Clear liquid
Refractive index
1.4400 to 1.4430 (20°C, 589 nm)
GC
>=98.5 %
Color scale
=<50 APHA
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text