Thermo Scientific Chemicals

Bis(trimethylsilyl)carbodiimide, 98%, Thermo Scientific Chemicals

Catalog number: 375130250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Bis(trimethylsilyl)carbodiimide, 98%, Thermo Scientific Chemicals

Catalog number: 375130250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 375130250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1000-70-0
IUPAC Name
2,2,6,6-tetramethyl-3,5-diaza-2,6-disilahepta-3,4-diene
Molecular Formula
C7H18N2Si2
InChI Key
KSVMTHKYDGMXFJ-UHFFFAOYSA-N
SMILES
C[Si](C)(C)N=C=N[Si](C)(C)C
Appearance (Form)
Liquid
Refractive index
1.4330 to 1.4350 (20°C, 589 nm)
GC
>=97.5 %
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text