Thermo Scientific Chemicals

Titanium(III) chloride tetrahydrofuran complex, 97%, Thermo Scientific Chemicals

Catalog number: 375160010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Titanium(III) chloride tetrahydrofuran complex, 97%, Thermo Scientific Chemicals

Catalog number: 375160010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 375160010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18039-90-2
IUPAC Name
tris(oxolane); trichlorotitanium
Molecular Formula
C12H24Cl3O3Ti
InChI Key
MIDYWIPTCYRMQF-UHFFFAOYSA-K
SMILES
Cl[Ti](Cl)Cl.C1CCOC1.C1CCOC1.C1CCOC1
Appearance (Color)
Blue-green to blue
Titration Argentometric
>=96.0 %
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text