Titanium(III) chloride tetrahydrofuran complex, 97%, Thermo Scientific Chemicals
Titanium(III) chloride tetrahydrofuran complex, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium(III) chloride tetrahydrofuran complex, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
View all
Glass Bottle
Catalog number 375160100
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
186.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
186.00 
Save 27.35 (15%)
Add to cart
Titanium(III) chloride tetrahydrofuran complex, 97%, Thermo Scientific Chemicals
Catalog number375160100
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
186.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
CAS18039-90-2
IUPAC Nametris(oxolane); trichlorotitanium
Molecular FormulaC12H24Cl3O3Ti
InChI KeyMIDYWIPTCYRMQF-UHFFFAOYSA-K
SMILESCl[Ti](Cl)Cl.C1CCOC1.C1CCOC1.C1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue-green to blue
Titration Argentometric>=96.0 %
Appearance (Form)Powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only