Blocker™ BSA
Blocker™ BSA
Thermo Scientific™

Blocker™ BSA

Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.
Promo ImagePromotion
Promo code:RBLOT25
Save up to 24% on Western Blot Reagents and Gels
From now until June 30th, get 16% off when you spend $400 or more, and get 24% off when you spend $750 or more on select western blot reagent and protein gels.Learn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBuffer
37520Promo ImageTBS
37525X3PBS
37525Promo ImagePBS
Catalog number 37520
Price (USD)
192.65
Online Exclusive
216.00
Save 23.35 (11%)
Each
Add to cart
Buffer:
TBS
Request bulk or custom format
Price (USD)
192.65
Online Exclusive
216.00
Save 23.35 (11%)
Each
Add to cart
Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.
Blocker BSA is a 10% (w/v) solution of high-quality BSA that is useful for saturating excess protein-binding sites on membranes and microplates in immunoassays. It is most frequently diluted 10-fold (to 1% BSA) in 1X PBS or 1X TBS for initial testing. Blocker BSA is usually more effective than nonfat milk for biotin-avidin systems because it contains a single purified protein that is devoid of endogenous biotin.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Blocking AgentSingle purified protein
BufferTBS
Concentration10% BSA
DescriptionBlocker BSA in TBS (10X)
For Use With (Application)Western Blot, ELISA, Immunohistochemistry (IHC)
FormLiquid
Product LineBlocker™
Product TypeBlocker BSA
Quantity125 mL
Unit SizeEach
Contents & Storage
Store at 4°C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link