tert-Butyl methyl ether, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl methyl ether, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 375211000
Price (USD)/ Each
57.65
Online exclusive
63.40 
Save 5.75 (9%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
57.65
Online exclusive
63.40 
Save 5.75 (9%)
Add to cart
tert-Butyl methyl ether, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Catalog number375211000
Price (USD)/ Each
57.65
Online exclusive
63.40 
Save 5.75 (9%)
-
Add to cart
Chemical Identifiers
CAS1634-04-4
IUPAC Name2-methoxy-2-methylpropane
Molecular FormulaC5H12O
InChI KeyBZLVMXJERCGZMT-UHFFFAOYSA-N
SMILESCOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Residue after evaporation=<0.002 %
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only