tert-Butyl methyl ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
tert-Butyl methyl ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl methyl ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 375220010
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
Add to cart
tert-Butyl methyl ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
Catalog number375220010
Price (USD)/ Each
97.65
Online exclusive
108.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1634-04-4
IUPAC Name2-methoxy-2-methylpropane
Molecular FormulaC5H12O
InChI KeyBZLVMXJERCGZMT-UHFFFAOYSA-N
SMILESCOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Water=<0.005 %
Peroxides=<0.005 % (as H2O2)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only