Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Blocker BSA in PBS (10X) |
Product Type | Blocker BSA |
Quantity | 3 x 200 mL |
Form | Liquid |
For Use With (Application) | Western Blot, ELISA, Immunohistochemistry (IHC) |
Product Line | Blocker™ |
Concentration | 10% BSA |
Buffer | PBS |
Blocking Agent | Single purified protein |
Unit Size | 3 x 200 mL |
Description | Blocker BSA in TBS (10X) |
Product Type | Blocker BSA |
Quantity | 125 mL |
Form | Liquid |
For Use With (Application) | Western Blot, ELISA, Immunohistochemistry (IHC) |
Product Line | Blocker™ |
Concentration | 10% BSA |
Buffer | TBS |
Blocking Agent | Single purified protein |
Unit Size | 125 mL |
Description | Blocker BSA in PBS (10X) |
Product Type | Blocker BSA |
Quantity | 200 mL |
Form | Liquid |
For Use With (Application) | Western Blot, ELISA, Immunohistochemistry (IHC) |
Product Line | Blocker™ |
Concentration | 10% BSA |
Buffer | PBS |
Blocking Agent | Single purified protein |
Unit Size | 200 mL |
Catalog Number | Specifications | Unit Size | Buffer | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
37525X3 | 3 x 200 mL | PBS | 480.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37520 | 125 mL | TBS | 183.65 Online price 199.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
37525 | 200 mL | PBS | 197.65 Online price 214.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|