StartingBlock™ Blocking Buffer
StartingBlock™ Blocking Buffer
Thermo Scientific™

StartingBlock™ Blocking Buffer

StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
37543StartingBlock T20 (TBS) Blocking Buffer, 1 L
37578StartingBlock (PBS) Blocking Buffer, 100 mL
37538StartingBlock (PBS) Blocking Buffer, 1 L
37579StartingBlock (TBS) Blocking Buffer, 100 mL
37538X3StartingBlock (PBS) Blocking Buffer, 3 x 1 L
37542StartingBlock (TBS) Blocking Buffer, 1 L
37539StartingBlock T20 (PBS) Blocking Buffer, 1 L
7 Options
Catalog number 37543
Price (USD)/ 1 L
248.65
Online exclusive
270.00 
Save 21.35 (8%)
-
Add to cart
Description:
StartingBlock T20 (TBS) Blocking Buffer, 1 L
Request bulk or custom format
Price (USD)/ 1 L
248.65
Online exclusive
270.00 
Save 21.35 (8%)
Add to cart
StartingBlock™ Blocking Buffer
Catalog number37543
Price (USD)/ 1 L
248.65
Online exclusive
270.00 
Save 21.35 (8%)
-
Add to cart
StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays. It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock Blocking Buffer is available in PBS and TBS, with and without Tween 20.
Features of StartingBlock Blocking Buffer include:
Strip and re-probe without re-blocking—blots stay blocked even after stripping with Restore Stripping Buffer (Cat. No. 21059)
Compatible with many detection systems—western blot, ELISA, and IHC with antibody or avidin/biotin probes (blocker is serum- and biotin-free)
Short blocking times—less than 15 minutes for nitrocellulose or PVDF membranes; almost instantaneous for polystyrene microplate wells

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionStartingBlock T20 (TBS) Blocking Buffer, 1 L
Product TypeBlocking Buffer
Quantity1 L
FormLiquid
For Use With (Application)Western Blot, ELISA
Product LineStartingBlock™
FormulationProprietary protein formulation in phosphate-buffered saline, pH 7.5, with 0.05% Tween 20
Concentration1X
BufferTBST
Blocking AgentSingle purified protein
Unit Size1 L
Contents & Storage
Upon receipt store at 4°C.