Thermo Scientific™

North2South™ Hybridization Buffer

Catalog number: 37549
Have Questions?
Thermo Scientific™

North2South™ Hybridization Buffer

Catalog number: 37549
Have Questions?
Catalog Number
37549
Unit Size
125 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
37549125 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer
Hybridization Buffer
Detection Method
Chromogenic
Product Line
North2South™
Product Type
Hybridization Buffer
Quantity
125 mL

Contents & Storage

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text